Novus Biologicals
Manufacturer Code:NBP220964
Catalog # NBP220964
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human ZNF148. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BERF-1 BFCOL1 CACCC box-binding protein CLL-associated antigen KW-10 HT-BETA pHZ-52 Transcription factor ZBP-89 ZBP89 ZBP-89 ZFP148zinc finger protein 148 (pHZ-52) Zinc finger DNA-binding protein 89 zinc finger protein 148; CACCC box-binding protein; CLL-associated antigen KW-10; Transcription factor ZBP-89; Zinc finger DNA-binding protein 89; Zinc finger protein 148
Gene Aliases: BERF-1; BFCOL1; HT-BETA; pHZ-52; ZBP-89; ZBP89; ZFP148; ZNF148
UniProt ID: (Human) Q9UQR1
Entrez Gene ID: (Human) 7707
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.