Novus Biologicals
Manufacturer Code:NBP29351920UL
Catalog # NB028915
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 100-180 of human ZNF134 (NP_003426.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC138499 MGC141970 pHZ-15 zinc finger protein 134 zinc finger protein 134 (clone pHZ-15); Zinc finger protein 134; zinc finger protein 134 (clone pHZ-15)
Gene Aliases: pHZ-15; ZNF134
UniProt ID: (Human) P52741
Entrez Gene ID: (Human) 7693
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.