Novus Biologicals
Manufacturer Code:NBP286909
Catalog # NB034531
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human ZMYND11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenovirus 5 E1A binding protein Adenovirus 5 E1A-binding protein bone morphogenetic protein receptor-associated molecule 1 BRAM1 MGC111056 MYND domain containing 11 Protein BS69 zinc finger MYND domain-containing protein 11 zinc finger MYND-type containing 11; Adenovirus 5 E1A-binding protein; Bone morphogenetic protein receptor-associated molecule 1; Protein BS69; Zinc finger MYND domain-containing protein 11; zinc finger, MYND-type containing 11
Gene Aliases: BRAM1; BS69; MRD30; ZMYND11
UniProt ID: (Human) Q15326
Entrez Gene ID: (Human) 10771
Molecular Function:
ligase
transcription cofactor
transcription factor
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.