Invitrogen
Manufacturer Code:PA566658
Catalog # PIPA566658
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human ZMYM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RP11-248I9.4 MYM RP11-181E22.4 zinc finger MYM-type protein 1 zinc finger MYM domain containing 1; Zinc finger MYM-type protein 1; zinc finger, MYM domain containing 1; zinc finger, MYM-type 1
Gene Aliases: MYM; ZMYM1
UniProt ID: (Human) Q5SVZ6
Entrez Gene ID: (Human) 79830
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.