Invitrogen
Manufacturer Code:PA513219
Catalog # PIPA513219
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1038-1067 amino acids from the C-terminal region of human RAI17 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ00092 protein; PIAS-like protein Zimp10; retinoic acid induced 17; Retinoic acid-induced protein 17; RP11-519K18.1; Zinc finger MIZ domain-containing protein 1; zinc finger, MIZ-type containing 1; zinc finger-containing, Miz1, PIAS-like protein on chromosome 10
Gene Aliases: hZIMP10; KIAA1224; MIZ; RAI17; TRAFIP10; ZIMP10; ZMIZ1
UniProt ID: (Human) Q9ULJ6
Entrez Gene ID: (Human) 57178
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.