Proteintech
Manufacturer Code:194291AP150UL
Catalog # 50-173-2402
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZIP7 Fusion Protein Ag13762 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H2-K region expressed gene 4; Histidine-rich membrane protein; Histidine-rich membrane protein Ke4; HLA class II region expressed gene KE4; Ke4 gene, mouse, human homolog of; KE4 RING5 SLC39A7 Zinc transporter SLC39A7 ZIP 7 ZIP7; Really interesting new gene 5 protein; RT1 class I, locus Ke4; RT1 region expressed gene KE4; solute carrier family 39 (zinc transporter), member 7; Solute carrier family 39 member 7; Zinc transporter SLC39A7; ZIP7; Zrt-, Irt-like protein 7
Gene Aliases: AA408174; AI117660; AL024048; D6S115E; D6S2244E; H-2Ke4; H2-KE4; HKE4; Ke-4; KE4; RING5; RT1-Ke4; Rt1Ke4; SLC39A7; ZIP7
UniProt ID: (Human) Q92504, (Mouse) Q31125
Entrez Gene ID: (Human) 7922, (Mouse) 14977, (Rat) 294281
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.