Invitrogen
Manufacturer Code:PA5101971
Catalog # PIPA5101971
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC39A4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1600025H15Rik Activated in W/Wv mouse stomach 2 AEZ AU041686 AWMS2 EMBL:AAI27515.1} mAWMS2 Slc39a4 slc39a4 {ECO:0000312 solute carrier family 39 (zinc transporter) member 4 solute carrier family 39 member 4 Zinc transporter ZIP4 ZIP4 ZIP-4 Zrt- and Irt-like protein 4; solute carrier family 39 (zinc transporter), member 4; Solute carrier family 39 member 4; Zinc transporter ZIP4; ZIP-4; Zrt- and Irt-like protein 4
Gene Aliases: AEZ; AWMS2; SLC39A4; ZIP4
UniProt ID: (Human) Q6P5W5
Entrez Gene ID: (Human) 55630
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.