Invitrogen
Manufacturer Code:PA534202
Catalog # PIPA534202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 15 amino acid peptide from internal region of human SLC39A14. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LIV-1 subfamily of ZIP zinc transporter 4; LZT-Hs4; Metal cation symporter ZIP14; SLC39A14 KIAA0062 LZT-Hs4 NET34 Zrt- Irt-like protein 14 Zinc transporter ZIP14 ZIP-14 ZIP14 Zrt- and Irt-like protein 14 Cig19; solute carrier family 39 (metal ion transporter), member 14; solute carrier family 39 (zinc transporter), member 14; Solute carrier family 39 member 14; ZIP-14; Zrt- and Irt-like protein 14; Zrt-, Irt-like protein 14
Gene Aliases: cig19; HMNDYT2; KIAA0062; LZT-Hs4; NET34; SLC39A14; ZIP14
UniProt ID: (Human) Q15043
Entrez Gene ID: (Human) 23516
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.