Invitrogen
Manufacturer Code:PA590883
Catalog # PIPA590883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SLC39A12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LIV-1 subfamily of ZIP zinc transporter 8; LZT-Hs8; solute carrier family 39 (metal ion transporter), member 12; solute carrier family 39 (zinc transporter), member 12; Solute carrier family 39 member 12; Zinc transporter ZIP12; ZIP-12; Zrt- and Irt-like protein 12
Gene Aliases: AW046938; bA570F3.1; Gm731; LZT-Hs8; SLC39A12; ZIP-12; ZIP12
UniProt ID: (Human) Q504Y0, (Mouse) Q5FWH7
Entrez Gene ID: (Human) 221074, (Mouse) 277468, (Rat) 291328
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.