Invitrogen
Manufacturer Code:PA521075
Catalog # PIPA521075
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide near the amino terminus of human ZIP12. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LIV-1 subfamily of ZIP zinc transporter 8; LZT-Hs8; LZT-Hs8 ZIP-12 bA570F3.1 LIV-1 subfamily of ZIP zinc transporter 8 solute carrier family 39 (metal ion transporter) member 12 solute carrier family 39 member 12 zinc transporter ZIP12 zrt- and Irt-like protein 12; solute carrier family 39 (metal ion transporter), member 12; solute carrier family 39 (zinc transporter), member 12; Solute carrier family 39 member 12; Zinc transporter ZIP12; ZIP-12; Zrt- and Irt-like protein 12
Gene Aliases: AW046938; bA570F3.1; Gm731; LZT-Hs8; SLC39A12; ZIP-12; ZIP12
UniProt ID: (Human) Q504Y0, (Mouse) Q5FWH7
Entrez Gene ID: (Human) 221074, (Mouse) 277468
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.