Invitrogen
Manufacturer Code:PA520678
Catalog # PIPA520678
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide near the center of the human ZIP10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LZT-Hs2 ZIP-10 solute carrier family 39 (metal ion transporter) member 10 solute carrier family 39 member 10 zinc transporter ZIP10 zrt- and Irt-like protein 10; solute carrier family 39 (metal ion transporter), member 10; solute carrier family 39 (zinc transporter), member 10; Solute carrier family 39 member 10; Zinc transporter ZIP10; ZIP-10; Zrt- and Irt-like protein 10
Gene Aliases: 2900042E17Rik; 5430433I10; KIAA1265; LZT-Hs2; mKIAA1265; SLC39A10; ZIP10
UniProt ID: (Human) Q9ULF5, (Mouse) Q6P5F6
Entrez Gene ID: (Human) 57181, (Mouse) 227059, (Rat) 363229
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.