Invitrogen
Manufacturer Code:PA577766
Catalog # PIPA577766
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)GEPELLVWRPEAVASE corresponding to amino acid residues 5- 20 of human ZIP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mZIP1; solute carrier family 39 (zinc transporter), member 1; solute carrier family 39 (zinc transporter), member 3; Solute carrier family 39 member 1; Zinc transporter 1 ZIP1 Zinc/Iron Regulated Transporter-Like SLC39A1; Zinc transporter ZIP1; Zinc-iron-regulated transporter-like; ZIP-1; Zrt- and Irt-like protein 1; Zrt/Irt-like protein 1
Gene Aliases: CGI-08; CGI-71; IRT1; SLC39A1; ZIP1; ZIRTL
UniProt ID: (Human) Q9Y380, (Mouse) Q9QZ03
Entrez Gene ID: (Human) 27173, (Mouse) 30791, (Rat) 361986
Molecular Function:
transmembrane receptor regulatory/adaptor protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.