Invitrogen
Manufacturer Code:MA520240
Catalog # PIMA520240
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G1 |
Immunogen | ZIC3 (NP&lowbar003404.1 182 a.a. &bsim 274 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: heterotaxy 1; Zic family member 3 (odd-paired homolog, Drosophila); Zinc finger protein 203; Zinc finger protein of the cerebellum 3; Zinc finger protein ZIC 1 Zinc finger protein 201 Zinc finger protein of the cerebellum 1 ZIC1 ZIC ZNF201 Zinc finger protein ZIC 2 Zinc finger protein of the cerebellum 2 ZIC2 Zinc finger protein ZIC 3 Zinc finger protein 203 Zinc finger protein of the cerebellum 3 ZIC3 ZNF203; Zinc finger protein ZIC 3
Gene Aliases: HTX; HTX1; VACTERLX; ZIC3; ZNF203
UniProt ID: (Human) O60481
Entrez Gene ID: (Human) 7547
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.