Novus Biologicals
Manufacturer Code:NBP133207
Catalog # NBP133207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ZIC3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: heterotaxy 1; heterotaxy 1 HTXHTX1 Zic family member 3 (odd-paired homolog Drosophila) Zinc finger protein 203 zinc finger protein ZIC 3 ZNF203Zinc finger protein of the cerebellum 3; Zic family member 3 (odd-paired homolog, Drosophila); Zinc finger protein 203; Zinc finger protein of the cerebellum 3; Zinc finger protein ZIC 3
Gene Aliases: HTX; HTX1; VACTERLX; ZIC3; ZNF203
UniProt ID: (Human) O60481
Entrez Gene ID: (Human) 7547
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.