Invitrogen
Manufacturer Code:PA538538
Catalog # PIPA538538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human ZIC1/2/3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GENA 29; heterotaxy 1; kumba; odd-paired homolog; Zic family member 1 (odd-paired homolog, Drosophila); Zic family member 2 (odd-paired homolog, Drosophila); Zic family member 3 (odd-paired homolog, Drosophila); Zinc finger protein 201; Zinc finger protein 203; Zinc finger protein of the cerebellum 1; Zinc finger protein of the cerebellum 2; Zinc finger protein of the cerebellum 3; Zinc finger protein ZIC 1; Zinc finger protein ZIC 1 Zinc finger protein 201 Zinc finger protein of the cerebellum 1 ZIC1 ZIC ZNF201 Zinc finger protein ZIC 2 Zinc finger protein of the cerebellum 2 ZIC2 Zinc finger protein ZIC 3 Zinc finger protein 203 Zinc finger protein of the cerebellum 3 ZIC3 ZNF203; Zinc finger protein ZIC 2; Zinc finger protein ZIC 3
Gene Aliases: Bn; CRS6; HPE5; HTX; HTX1; Ka; Ku; VACTERLX; ZIC; ZIC1; ZIC2; ZIC3; ZNF201; ZNF203
UniProt ID: (Human) Q15915, (Human) O95409, (Human) O60481, (Mouse) P46684, (Mouse) Q62521
Entrez Gene ID: (Human) 7545, (Human) 7546, (Human) 7547, (Mouse) 22771, (Mouse) 22772, (Mouse) 22773
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.