Invitrogen
Manufacturer Code:PA529094
Catalog # PIPA529094
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 280 and 564 of Human FOG2/ZFPM2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DIH3 FOG2 ZNF89B hFOG-2 FOG-2 Friend of GATA2 friend of GATA 2 friend of GATA protein 2 transcription factor GATA4 modulator of zinc finger protein 89B zinc finger protein ZFPM2; FOG-2; Friend of GATA 2; Friend of GATA protein 2; friend of GATA-1 2; friend of GATA-2; Friend of GATA2; mFOG-2; transcription factor GATA4, modulator of; Zinc finger protein 89B; Zinc finger protein multitype 2; Zinc finger protein ZFPM2; zinc finger protein, multitype 2
Gene Aliases: B330005D23Rik; DIH3; FOG-2; FOG2; hFOG-2; SRXY9; ZC2HC11B; ZFPM2; ZNF89B
UniProt ID: (Human) Q8WW38, (Mouse) Q8CCH7
Entrez Gene ID: (Human) 23414, (Mouse) 22762, (Rat) 314930
Molecular Function:
nucleic acid binding
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.