Novus Biologicals
Manufacturer Code:NBP17941120UL
Catalog # NBP179420UL
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ZFP90The immunogen for this antibody is ZFP90. Peptide sequence SSLVQHQRIHTGEKPYRCNLCGRSFRHGTSLTQHEVTHSGEKPFQCKECG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FOXP3-interacting KRAB domain-containing protein; KIAA1954 NK10 zfp-90 zinc finger protein 476 zinc finger protein 756 zinc finger protein 90 homolog zinc finger protein 90 homolog (mouse) ZNF756; Zfp-90; zinc finger protein 476; Zinc finger protein 756; Zinc finger protein 90 homolog
Gene Aliases: FIK; KIAA1954; NK10; zfp-90; ZFP90; ZNF756
UniProt ID: (Human) Q8TF47
Entrez Gene ID: (Human) 146198
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.