Abnova
Manufacturer Code:H00000677M02
Catalog # 89-016-366
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A3 |
Immunogen | ZFP36L1 (NP_004917 1 a.a. ~ 108 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ZFP36L1.
For Research Use Only
Protein Aliases: Berg36; BRF1; Butyrate response factor 1; cMG1; early response factor Berg36; EGF-response factor 1; ERF-1; ERF1; mRNA decay activator protein ZFP36L1; RNF162B; TIS11B; TPA-induced sequence 11b; ZFP36 ring finger protein-like 1; ZFP36-like 1; zinc finger protein 36, C3H type-like 1; Zinc finger protein 36, C3H1 type-like 1; zinc finger protein, C3H type, 36-like 1
Gene Aliases: BERG36; BRF1; cMG1; ERF-1; ERF1; RNF162B; TIS11B; ZFP36L1
UniProt ID: (Human) Q07352
Entrez Gene ID: (Human) 677
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.