Novus Biologicals
Manufacturer Code:NBP286904
Catalog # NB034526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human ZFP36L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Butyrate response factor 1; early response factor Berg36; EGF-response factor 1; ERF-1; mRNA decay activator protein ZFP36L1; TPA-induced sequence 11b; ZBRK1KRAB zinc finger protein ZFQR ZFQR Zinc finger and BRCA1-interacting protein with a KRAB domain 1 zinc finger protein 350 Zinc finger protein ZBRK1 zinc-finger protein ZBRK1; ZFP36 ring finger protein-like 1; ZFP36-like 1; zinc finger protein 36, C3H type-like 1; Zinc finger protein 36, C3H1 type-like 1; zinc finger protein, C3H type, 36-like 1
Gene Aliases: BERG36; BRF1; cMG1; ERF-1; ERF1; RNF162B; TIS11B; ZFP36L1
UniProt ID: (Human) Q07352
Entrez Gene ID: (Human) 677
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.