OriGene
Manufacturer Code:CF502385
Catalog # 50-166-1294
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4E1 |
Immunogen | Full length human recombit protein of human ZFP36 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: G0/G1 switch regulatory protein 24; G0S24 GOS24 NUP475 RNF162A TIS11 TTP zfp-36; Growth factor-inducible nuclear protein NUP475; mRNA decay activator protein ZFP36; Tristetraprolin; tristetraproline; Zfp-36; Zinc finger protein 36; zinc finger protein 36 homolog; zinc finger protein 36, C3H type, homolog; zinc finger protein, C3H type, 36 homolog
Gene Aliases: G0S24; GOS24; NUP475; RNF162A; TIS11; TIS11A; TTP; zfp-36; ZFP36
UniProt ID: (Human) P26651
Entrez Gene ID: (Human) 7538
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.