Invitrogen
Manufacturer Code:PA541450
Catalog # PIPA541450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ZFP36 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G0/G1 switch regulatory protein 24; Growth factor-inducible nuclear protein NUP475; mRNA decay activator protein ZFP36; Tax interacting protein; TNF alpha inhibitor; TPA-induced sequence 11; Tristetraprolin; tristetraproline; TTP; Zfp-36; Zinc finger protein 36; Zinc finger protein 36 C3H type homolog (mouse) G0S24 GOS24 NUP475 RNF162A TIS11 TTP; zinc finger protein 36 homolog; zinc finger protein 36, C3H type, homolog; zinc finger protein, C3H type, 36 homolog
Gene Aliases: G0S24; GOS24; NUP475; RNF162A; TIS11; TIS11A; TIS11D; TISII; TTP; Zfp-36; ZFP36
UniProt ID: (Sheep) Q6S9E0, (Human) P26651, (Bovine) P53781, (Mouse) P22893, (Rat) P47973
Entrez Gene ID: (Sheep) 443283, (Human) 7538, (Dog) 484510, (Rabbit) 100340391, (Bovine) 282127, (Guinea pig) 100733110, (Mouse) 22695, (Rat) 79426
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.