Abnova
Manufacturer Code:H00000463M01
Catalog # 89-016-232
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3B1 |
Immunogen | ZFHX3 (NP_008816 2811 a.a. ~ 2910 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ZFHX3.
For Research Use Only
Protein Aliases: alpha-fetoprotein enhancer binding protein; Alpha-fetoprotein enhancer-binding protein; AT motif-binding factor 1; AT-binding transcription factor 1; ATBF1; ATBT; ZFH-3; Zinc finger homeobox protein 3; Zinc finger homeodomain protein 3
Gene Aliases: ATBF1; ATBT; C16orf47; ZFHX3; ZNF927
UniProt ID: (Human) Q15911
Entrez Gene ID: (Human) 463
Molecular Function: RNA binding protein actin family cytoskeletal protein cytoskeletal protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.