Invitrogen
Manufacturer Code:PA544979
Catalog # PIPA544979
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ZER1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hzyg; Protein zer-1 homolog; zer-1 homolog; ZYG homolog; Zyg-11 homolog B-like protein; zyg-11 related, cell cycle regulator; Zyg11b-like protein
Gene Aliases: C230075L19Rik; C9orf60; RGD1310284; ZER1; ZYG; ZYG11BL
UniProt ID: (Human) Q7Z7L7, (Mouse) Q80ZJ6
Entrez Gene ID: (Rabbit) 100350464, (Horse) 100066859, (Dog) 491312, (Bovine) 540351, (Human) 10444, (Rat) 311842, (Mouse) 227693, (Guinea pig) 100729084
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.