Invitrogen
Manufacturer Code:PA521807
Catalog # PIPA521807
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 537 and 766 of ZER1 (Uniprot IDNo.Q7Z7L7) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C9orf60 Hzyg RP11-545E17.4 ZYG ZYG11BL ZYG homolog homolog of Zyg-11 protein zer-1 homolog zyg-11 homolog B-like protein; Hzyg; Protein zer-1 homolog; zer-1 homolog; ZYG homolog; Zyg-11 homolog B-like protein; zyg-11 related, cell cycle regulator; Zyg11b-like protein
Gene Aliases: C230075L19Rik; C9orf60; ZER1; ZYG; ZYG11BL
UniProt ID: (Human) Q7Z7L7, (Mouse) Q80ZJ6
Entrez Gene ID: (Human) 10444, (Mouse) 227693
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.