R&D Systems
Manufacturer Code:MAB73782SP
Catalog # MAB73782SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 923328 |
Immunogen | <em>E. coli</em>-derived recombinant human ZEB2/SIP1 Asn363-Lys537 Accession # O60315 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HSPC082 KIAA0569FLJ42816 SIP1 SIP1HSPC082 Smad-interacting protein 1 SMADIP1 SMADIP1ZFHX1BSIP-1 ZFHX1B ZFX1B zinc finger E-box binding homeobox 2 zinc finger E-box-binding homeobox 2 zinc finger homeobox 1b Zinc finger homeobox protein 1b; SMAD interacting protein 1; Smad-interacting protein 1; SMADIP1; Zinc finger E-box-binding homeobox 2; zinc finger homeobox 1b; Zinc finger homeobox protein 1b
Gene Aliases: HRIHFB2411; HSPC082; KIAA0569; SIP-1; SIP1; SMADIP1; ZEB2; ZFHX1B; ZFX1B
UniProt ID: (Human) O60315
Entrez Gene ID: (Human) 9839
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.