Invitrogen
Manufacturer Code:PA540759
Catalog # PIPA540759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ZEB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SMAD interacting protein 1; Smad-interacting protein 1; SMADIP1; Zinc finger E-box binding homeobox 2 KIAA0569 SIP-1 SIP1 SMADIP1 ZFHX1B HSPC082; Zinc finger E-box-binding homeobox 2; zinc finger homeobox 1b; Zinc finger homeobox protein 1b
Gene Aliases: 9130203F04Rik; D130016B08Rik; HRIHFB2411; HSPC082; KIAA0569; SIP-1; SIP1; SMADIP1; ZEB2; ZFHX1B; ZFX1B; Zfxh1b
UniProt ID: (Human) O60315, (Mouse) Q9R0G7
Entrez Gene ID: (Dog) 483909, (Human) 9839, (Horse) 100057772, (Bovine) 539873, (Rabbit) 100344650, (Pig) 100739669, (Mouse) 24136, (Guinea pig) 100727818, (Rat) 311071
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.