OriGene
Manufacturer Code:CF802313
Catalog # 50-166-5939
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7E12 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-350 of human ZEB1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AREB6 BZP DELTAEF1 FECD6 NIL2A PPCD3 TCF8 ZFHEP ZFHX1A; delta-crystallin enhancer binding factor 1; Negative regulator of IL2; NIL-2-A zinc finger protein; posterior polymorphous corneal dystrophy 3; TCF-8; Transcription factor 8; transcription factor 8 (represses interleukin 2 expression); Zinc finger E-box-binding homeobox 1; zinc finger homeodomain enhancer-binding protein
Gene Aliases: AREB6; BZP; DELTAEF1; FECD6; NIL2A; PPCD3; TCF8; ZEB1; ZFHEP; ZFHX1A
UniProt ID: (Human) P37275
Entrez Gene ID: (Human) 6935
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.