Invitrogen
Manufacturer Code:PA520979
Catalog # PIPA520979
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide near the center of human ZEB1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: delta-crystallin enhancer binding factor 1; Negative regulator of IL2; NIL-2-A zinc finger protein; posterior polymorphous corneal dystrophy 3; RP11-472N13.4 AREB6 BZP DELTAEF1 FECD6 NIL2A PPCD3 TCF8 ZFHEP ZFHX1A delta-crystallin enhancer binding factor 1 negative regulator of IL2 posterior polymorphous corneal dystrophy 3 transcription factor 8 (represses interleukin 2 expression) zinc finger E-box-binding homeobox 1 zinc finger homeodomain enhancer-binding protein; TCF-8; Transcription factor 8; transcription factor 8 (represses interleukin 2 expression); Zinc finger E-box-binding homeobox 1; zinc finger homeodomain enhancer-binding protein
Gene Aliases: AREB6; BZP; DELTAEF1; FECD6; NIL2A; PPCD3; TCF8; ZEB1; ZFHEP; ZFHX1A
UniProt ID: (Human) P37275
Entrez Gene ID: (Human) 6935
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.