Novus Biologicals
Manufacturer Code:NBP16893020UL
Catalog # NBP16893020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ZEB1 (zinc finger E-box binding homeobox 1) The peptide sequence was selected from the N terminal of ZEB1. Peptide sequence NPRRNNVTNYNTVVETNSDSDDEDKLHIVEEESVTDAADCEGVPEDDLPT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AREB6MGC133261 delta-crystallin enhancer binding factor 1 DELTAEF1 FECD6 Negative regulator of IL2 NIL2A NIL-2-A NIL-2-A zinc finger protein posterior polymorphous corneal dystrophy 3 PPCD3 TCF-8 TCF8BZP Transcription factor 8 transcription factor 8 (represses interleukin 2 expression) ZEB Zfhep ZFHX1A zinc finger E-box binding homeobox 1 zinc finger E-box-binding homeobox 1 zinc finger homeodomain enhancer-binding protein; delta-crystallin enhancer binding factor 1; Negative regulator of IL2; NIL-2-A zinc finger protein; posterior polymorphous corneal dystrophy 3; TCF-8; Transcription factor 8; transcription factor 8 (represses interleukin 2 expression); Zinc finger E-box-binding homeobox 1; zinc finger homeodomain enhancer-binding protein
Gene Aliases: AREB6; BZP; DELTAEF1; FECD6; NIL2A; PPCD3; TCF8; ZEB1; ZFHEP; ZFHX1A
UniProt ID: (Human) P37275
Entrez Gene ID: (Human) 6935
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.