Proteintech
Manufacturer Code:215441AP150UL
Catalog # 50-173-3474
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZEB1 Fusion Protein Ag16204 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AREB6 BZP DELTA EF1 delta EF1 Negative regulator of IL2 NIL 2 A NIL 2 A zinc finger protein NIL 2A NIL2A TCF 8 TCF8 Transcription factor 8 ZEB ZEB1 ZFHEP ZFHX1A; Delta EF1; delta-crystallin enhancer binding factor 1; MEB1; Negative regulator of IL2; NIL-2-A zinc finger protein; posterior polymorphous corneal dystrophy 3; TCF-8; Transcription factor 8; transcription factor 8 (represses interleukin 2 expression); twirler; Zfhep; Zinc finger E-box-binding homeobox 1; Zinc finger homeobox protein 1a; Zinc finger homeodomain enhancer-binding protein
Gene Aliases: 3110032K11Rik; [delta]EF1; AREB6; BZP; DELTAEF1; FECD6; MEB1; Nil2; NIL2A; PPCD3; TCF-8; Tcf18; TCF8; Tw; ZEB; ZEB1; ZFHEP; ZFHX1A; Zfx1a; Zfx1ha
UniProt ID: (Human) P37275, (Mouse) Q64318, (Rat) Q62947
Entrez Gene ID: (Human) 6935, (Mouse) 21417, (Rat) 25705
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.