Invitrogen
Manufacturer Code:PA599617
Catalog # PIPA599617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DHHC-9 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6430508G22 9530098M12Rik antigen MMSA-1 Asp-His-His-Cys domain containing protein 9 CGI89 CGI-89 CXorf11 DHHC9 DHHC-9 membrane-associated DHHC9 zinc finger protein MMSA1 MRXSZ Palmitoyltransferase ZDHHC9 UNQ261/PRO298 ZDHHC10 ZDHHC9 zinc finger DHHC domain-containing protein 9 zinc finger DHHC-type containing 9 zinc finger protein 379 Zinc finger protein 380 zinc finger DHHC domain containing 10 zinc finger DHHC domain containing 9 zinc finger DHHC-type containing 9 ZNF379 ZNF380; antigen MMSA-1; Asp-His-His-Cys domain containing protein 9; DHHC-9; DHHC9; membrane-associated DHHC9 zinc finger protein; Palmitoyltransferase ZDHHC9; Zinc finger DHHC domain-containing protein 9; Zinc finger protein 379; Zinc finger protein 380; zinc finger, DHHC domain containing 10; zinc finger, DHHC domain containing 9; zinc finger, DHHC-type containing 9
Gene Aliases: 6430508G22; 9530098M12Rik; CGI-89; CGI89; CXorf11; DHHC9; MMSA1; MRXSZ; UNQ261/PRO298; ZDHHC10; ZDHHC9; ZNF379; ZNF380
UniProt ID: (Human) Q9Y397, (Mouse) P59268
Entrez Gene ID: (Human) 51114, (Rat) 302808, (Mouse) 208884
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.