Novus Biologicals
Manufacturer Code:NBP2837970.1ML
Catalog # NB031726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human ZDHHC9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antigen MMSA-1; Asp-His-His-Cys domain containing protein 9; CGI89 CGI-89 chromosome X open reading frame 11 DHHC-9 EC 2.3.1 EC 2.3.1.- EC 2.3.1.12 MRXSZ ZDHHC10 Zinc finger DHHC domain-containing protein 9 zinc finger DHHC-type containing 10 zinc finger DHHC-type containing 9 ZNF379MMSA1 ZNF380DHHC domain containing 10; DHHC-9; Palmitoyltransferase ZDHHC9; Zinc finger DHHC domain-containing protein 9; Zinc finger protein 379; Zinc finger protein 380; zinc finger, DHHC domain containing 10; zinc finger, DHHC-type containing 9
Gene Aliases: CGI-89; CGI89; CXorf11; DHHC9; MMSA1; MRXSZ; UNQ261/PRO298; ZDHHC10; ZDHHC9; ZNF379; ZNF380
UniProt ID: (Human) Q9Y397
Entrez Gene ID: (Human) 51114
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.