Invitrogen
Manufacturer Code:PA5110591
Catalog # PIPA5110591
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 240-380 of human ZDHHC8 (NP_037505.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D16H22S1738E DHHC8 DHHC-8 E330009O14Rik KIAA1292 Op53c05 Probable palmitoyltransferase ZDHHC8 ZDHHC8 ZDHHCL1 Zinc finger DHHC domain-containing protein 8 Zinc finger protein 378 ZNF378; DHHC-8; DHHC-containing protein 8; membrane-associated DHHC8 zinc finger protein; Palmitoyltransferase ZDHHC8; probable palmitoyltransferase ZDHHC8; Zinc finger DHHC domain-containing protein 8; Zinc finger protein 378; zinc finger, DHHC domain like containing 1; zinc finger, DHHC-type containing 8
Gene Aliases: D16H22S1738E; DHHC-8; DHHC8; E330009O14Rik; KIAA1292; Op53c05; ZDHHC8; ZDHHCL1; ZNF378
UniProt ID: (Human) Q9ULC8, (Mouse) Q5Y5T5
Entrez Gene ID: (Human) 29801, (Mouse) 27801
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.