Novus Biologicals
Manufacturer Code:NB10057849
Catalog # NB10057849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-QRDHPQLKTPPSK corresponding to internal region according to NP_037505.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DHHC-8; DHHC8 EC 2.3.1 EC 2.3.1.- KIAA1292DHHC-8 membrane-associated DHHC8 zinc finger protein probable palmitoyltransferase ZDHHC8 ZDHHCL1 Zinc finger DHHC domain-containing protein 8 Zinc finger protein 378 zinc finger DHHC-type containing 8 ZNF378zinc finger DHHC domain like containing 1; membrane-associated DHHC8 zinc finger protein; Palmitoyltransferase ZDHHC8; Zinc finger DHHC domain-containing protein 8; Zinc finger protein 378; zinc finger, DHHC domain like containing 1; zinc finger, DHHC-type containing 8
Gene Aliases: DHHC8; KIAA1292; ZDHHC8; ZDHHCL1; ZNF378
UniProt ID: (Human) Q9ULC8
Entrez Gene ID: (Human) 29801
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.