Abnova
Manufacturer Code:PAB7217
Catalog # 89-133-054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human ZDHHC8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of ZDHHC8.
For Research Use Only
Protein Aliases: DHHC-8; membrane-associated DHHC8 zinc finger protein; Palmitoyltransferase ZDHHC8; ZDHHCL1; Zinc finger DHHC domain-containing protein 8; Zinc finger protein 378; zinc finger, DHHC domain like containing 1; zinc finger, DHHC-type containing 8; ZNF378
Gene Aliases: DHHC8; KIAA1292; ZDHHC8; ZDHHCL1; ZNF378
UniProt ID: (Human) Q9ULC8
Entrez Gene ID: (Human) 29801
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.