Invitrogen
Manufacturer Code:PA5116795
Catalog # PIPA5116795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human ZDHHC7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyltransferase ZDHHC7; AL024087 DHHC7 DHHC-7 GABA-A receptor-associated membrane protein 2 Gramp2 membrane-associated DHHC7 zinc finger protein Palmitoyltransferase ZDHHC7 putative zinc finger protein SERZ-1 Sertoli cell gene with a zinc finger domain protein Sertoli cell gene with zinc finger domain-β Serz SERZ1 Serz-1 SERZ-B ZDHHC7 zinc finger DHHC domain-containing protein 7 zinc finger DHHC-type containing 7 zinc finger protein 370 zinc finger DHHC domain containing 7 zinc finger DHHC-type containing 7 ZNF370; DHHC-7; GABA-A receptor-associated membrane protein 2; Palmitoyltransferase ZDHHC7; Sertoli cell gene with zinc finger domain-β SERZ-beta; Zinc finger DHHC domain-containing protein 7; zinc finger protein 370; zinc finger, DHHC domain containing 7; zinc finger, DHHC-type containing 7
Gene Aliases: AL024087; DHHC7; Gramp2; SERZ-B; SERZ1; ZDHHC7; ZNF370
UniProt ID: (Human) Q9NXF8, (Mouse) Q91WU6
Entrez Gene ID: (Human) 55625, (Mouse) 102193
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.