Abnova
Manufacturer Code:H00055625B01P
Catalog # 89-017-125
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZDHHC7 (AAH17702 1 a.a. ~ 208 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human ZDHHC7 protein.
For Research Use Only
Protein Aliases: Acyltransferase ZDHHC7; DHHC-7; FLJ10792; FLJ20279; Palmitoyltransferase ZDHHC7; Sertoli cell gene with zinc finger domain-β Zinc finger DHHC domain-containing protein 7; zinc finger protein 370; zinc finger, DHHC domain containing 7; zinc finger, DHHC-type containing 7; ZNF370
Gene Aliases: DHHC7; SERZ-B; SERZ1; ZDHHC7; ZNF370
UniProt ID: (Human) Q9NXF8
Entrez Gene ID: (Human) 55625
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.