Invitrogen
Manufacturer Code:PA5113325
Catalog # PIPA5113325
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Palmitoyltransferase ZDHHC6 protein (227-409AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DHHC-6; DHHC-6 Palmitoyltransferase ZDHHC6 Transmembrane protein H4 ZDHHC6 Zinc finger DHHC domain-containing protein 6 zinc finger DHHC-type containing 6 Zinc finger protein 376 zinc finger DHHC-type containing 6 ZNF376; H4 homolog; Palmitoyltransferase ZDHHC6; probable palmitoyltransferase ZDHHC6; Stearoyltransferase ZDHHC6; Transmembrane protein H4; Zinc finger DHHC domain-containing protein 6; Zinc finger protein 376; zinc finger, DHHC-type containing 6
Gene Aliases: 2400007G07Rik; 5930409M18Rik; AI462345; C77369; DHHC-6; ZDHHC6; ZNF376
UniProt ID: (Human) Q9H6R6, (Mouse) Q9CPV7
Entrez Gene ID: (Human) 64429, (Mouse) 66980
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.