Invitrogen
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | 1:1,000 |
Host / Isotype | Rabbit / IgG |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 288-317 amino acids from the C-terminal region of human ZDHHC6. |
Conjugate | Unconjugated |
Form | Liquid |
Concentration | 0.4 mg/mL |
Storage conditions | -20° C, Avoid Freeze/Thaw Cycles |
RRID | AB_2634174 |
Predicted to react with bovine based on sequence homology.
Endoplasmic reticulum palmitoyl acyltransferase that mediates palmitoylation of proteins such as AMFR, CALX, ITPR1 and TFRC (PubMed:22314232, PubMed:22728137, PubMed:25368151, PubMed:28826475). Palmitoylates calnexin (CALX), which is required for its association with the ribosome-translocon complex and efficient folding of glycosylated proteins (PubMed:22314232). Mediates palmitoylation of AMFR, promoting AMFR distribution to the peripheral endoplasmic reticulum (PubMed:22728137). Together with SELENOK, palmitoylates ITPR1 in immune cells, leading to regulate ITPR1 stability and function (PubMed:25368151). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.