Novus Biologicals
Manufacturer Code:NBP2837960.1ML
Catalog # NB031725
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human ZDHHC5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DHHC domain containing 5 DKFZp586K0524 zinc finger DHHC-type containing 5 ZNF375; DHHC-5; membrane-associated DHHC5 zinc finger protein; Palmitoyltransferase ZDHHC5; probable palmitoyltransferase ZDHHC5; Zinc finger DHHC domain-containing protein 5; Zinc finger protein 375; zinc finger, DHHC domain containing 5; zinc finger, DHHC-type containing 5
Gene Aliases: DHHC5; KIAA1748; ZDHHC5; ZNF375
UniProt ID: (Human) Q9C0B5
Entrez Gene ID: (Human) 25921
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.