Abnova
Manufacturer Code:PAB20751
Catalog # 89-122-253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human ZDHHC5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant ZDHHC5.
For Research Use Only
Protein Aliases: DHHC-5; DKFZp586K0524; KIAA1748; membrane-associated DHHC5 zinc finger protein; Palmitoyltransferase ZDHHC5; probable palmitoyltransferase ZDHHC5; Zinc finger DHHC domain-containing protein 5; Zinc finger protein 375; zinc finger, DHHC domain containing 5; zinc finger, DHHC-type containing 5; ZNF375
Gene Aliases: DHHC5; KIAA1748; ZDHHC5; ZNF375
UniProt ID: (Human) Q9C0B5
Entrez Gene ID: (Human) 25921
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.