Invitrogen
Manufacturer Code:PA5113122
Catalog # PIPA5113122
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Palmitoyltransferase ZDHHC23 protein (187-302AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DHHC-23; DHHC-23 NIDD Palmitoyltransferase ZDHHC23 ZDHHC23 Zinc finger DHHC domain-containing protein 23 zinc finger DHHC-type containing 23; DHHC-containing protein 11; nNOS-intereacting DHHC-containing Dem protein-L; Palmitoyltransferase ZDHHC23; probable palmitoyltransferase ZDHHC23; Zinc finger DHHC domain-containing protein 23
Gene Aliases: DHHC-23; Gm1751; Gm779; NIDD; ZDHHC23
UniProt ID: (Human) Q8IYP9, (Mouse) Q5Y5T3
Entrez Gene ID: (Human) 254887, (Mouse) 332175
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.