Abnova
Manufacturer Code:PAB0263
Catalog # 89-114-085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to full length human ZDHHC23. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against full-length recombinant ZDHHC23.
For Research Use Only
Protein Aliases: DHHC-23; MGC42530; NIDD; Palmitoyltransferase ZDHHC23; Zinc finger DHHC domain-containing protein 23
Gene Aliases: DHHC-23; NIDD; ZDHHC23
UniProt ID: (Human) Q8IYP9
Entrez Gene ID: (Human) 254887
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.