Invitrogen
Manufacturer Code:PA570154
Catalog # PIPA570154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human ZDHHC21 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9130404H11Rik DHHC-21 DNZ1 HSPC097 probable palmitoyltransferase ZDHHC21 zinc finger DHHC domain-containing protein 21 zinc finger DHHC domain containing 21; DHHC domain-containing cysteine-rich protein 21; DHHC-21; DHHC21; GABA-A receptor-associated membrane protein 3; Palmitoyltransferase ZDHHC21; palmitoyltransferase ZDHHC21-like protein; probable palmitoyltransferase ZDHHC21; Zinc finger DHHC domain-containing protein 21; zinc finger DHHC-type containing 21; zinc finger, DHHC domain containing 21; zinc finger, DHHC-type containing 21
Gene Aliases: 9130404H11Rik; AL024349; D130004H04Rik; dep; DHHC-21; DHHC21; DNZ1; Gramp3; HSPC097; ZDHHC21
UniProt ID: (Human) Q8IVQ6, (Bovine) A2VDT6, (Mouse) Q9D270
Entrez Gene ID: (Rabbit) 100340626, (Human) 340481, (Dog) 481546, (Bovine) 535814, (Horse) 100062221, (Rat) 298184, (Mouse) 68268, (Guinea pig) 100723190
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.