Invitrogen
Manufacturer Code:PA5116793
Catalog # PIPA5116793
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human ZDHHC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5730415P04Rik 6430583A19Rik DHHC2 DHHC-2 membrane-associated DHHC2 zinc finger protein palmitoyltransferase ZDHHC2 REAM rec Reduced expression associated with metastasis protein reduced expression in cancer protein small rec srec testis tissue sperm-binding protein Li 56e Zdhhc2 Zinc finger DHHC domain-containing protein 2 zinc finger DHHC-type containing 2 zinc finger protein 372 zinc finger DHHC domain containing 2 zinc finger DHHC-type containing 2 ZNF372; Acyltransferase ZDHHC2; DHHC-2; membrane-associated DHHC2 zinc finger protein; Palmitoyltransferase ZDHHC2; Ream; Rec; Reduced expression associated with metastasis protein; Reduced expression in cancer protein; small rec; testis tissue sperm-binding protein Li 56e; Zinc finger DHHC domain-containing protein 2; Zinc finger protein 372; zinc finger, DHHC domain containing 2; zinc finger, DHHC-type containing 2
Gene Aliases: DHHC2; REAM; REC; Srec; ZDHHC2; ZNF372
UniProt ID: (Human) Q9UIJ5, (Rat) Q9JKR5
Entrez Gene ID: (Human) 51201, (Rat) 246326
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.