Proteintech
Manufacturer Code:216271AP150UL
Catalog # 50-173-3520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ZDHHC15 Fusion Protein Ag16152 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyltransferase ZDHHC15; DHHC 15 MRX91 Palmitoyltransferase ZDHHC15 ZDHHC15; DHHC-15; Palmitoyltransferase ZDHHC15; Zinc finger DHHC domain-containing protein 15; zinc finger, DHHC domain containing 15; zinc finger, DHHC-type containing 15
Gene Aliases: MRX91; UNQ1969/PRO4501; ZDHHC15
UniProt ID: (Human) Q96MV8
Entrez Gene ID: (Human) 158866
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.