Invitrogen
Manufacturer Code:PA569184
Catalog # PIPA569184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human ZDHHC14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DHHC domain-containing cysteine-rich protein 14; DHHC-14; membrane-associated DHHC14 zinc finger protein; NEW1 domain containing protein; NEW1 domain-containing protein; NEW1CP; NEW1CP DHHC-14 NEW1 domain containing protein NEW1 domain-containing protein probable palmitoyltransferase ZDHHC14 zinc finger DHHC domain-containing protein 14 zinc finger DHHC domain containing 14; Palmitoyltransferase ZDHHC14; probable palmitoyltransferase ZDHHC14; Zinc finger DHHC domain-containing protein 14; zinc finger DHHC-type containing 14; zinc finger, DHHC domain containing 14; zinc finger, DHHC-type containing 14
Gene Aliases: B530001K09Rik; Dhhc14; NEW1CP; si:ch211-199l3.1; ZDHHC14
UniProt ID: (Human) Q8IZN3, (Mouse) Q8BQQ1
Entrez Gene ID: (Dog) 612148, (Bovine) 517451, (Rabbit) 100337908, (Horse) 100051142, (Human) 79683, (Mouse) 224454, (Rat) 499014, (Guinea pig) 100715880, (Zebrafish) 569667
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.