Invitrogen
Manufacturer Code:MA522031
Catalog # PIMA522031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A4 |
Immunogen | ZDHHC13 (NP&lowbar061901.2 181 a.a. &bsim 280 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DHHC-13; HIP14-related protein; huntingtin interacting protein HIP3RP; Huntingtin-interacting protein 14-related protein; Huntingtin-interacting protein HIP3RP; Palmitoyltransferase ZDHHC13; probable palmitoyltransferase ZDHHC13; Putative MAPK-activating protein PM03; Putative NF-kappa-B-activating protein 209; ZDHHC13 HIP14L HIP3RP FLJ10852 FLJ10941 MGC64994 zinc finger DHHC domain containing 13 HIP14-like HIP14-related protein huntingtin interacting protein HIP3RP hypothetical protein similar to ankyrin repeat-containing priotein AKR1 zinc finger DHHC-type containing 13; Zinc finger DHHC domain-containing protein 13; zinc finger, DHHC domain containing 13; zinc finger, DHHC-type containing 13
Gene Aliases: HIP14L; HIP3RP; ZDHHC13
UniProt ID: (Human) Q8IUH4
Entrez Gene ID: (Human) 54503
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.