EMD Millipore
Manufacturer Code:MABT149825UL
Catalog # MABT149825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 26D1 |
Immunogen | MBP-tagged recombinant fragment corresponding to 111 amino acids from the N-terminal region of human Palmitoyltransferase ZDHHC13 isoform 2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Stable for 1 year at 2-8°C from date of receipt. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Palmitoyltransferase ZDHHC13EC: 2.3.1.22Huntingtin-interacting protein 14-related proteinHIP14-related proteinHuntingtin-interacting protein HIP3RPPutative MAPK-activating protein PM03Putative NF-kappa-B-activating protein 209Zinc finger DHHC domain-containing protein 13DHHC-13
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.