Invitrogen
Manufacturer Code:PA542565
Catalog # PIPA542565
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ZDHHC13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DHHC domain-containing protein 13; DHHC-13; HIP14-related protein; huntingtin interacting protein HIP3RP; Huntingtin-interacting protein 14-related protein; Huntingtin-interacting protein HIP3RP; Palmitoyltransferase ZDHHC13; probable palmitoyltransferase ZDHHC13; Putative MAPK-activating protein PM03; Putative NF-kappa-B-activating protein 209; Putative palmitoyltransferase ZDHHC13; Zinc finger DHHC domain-containing protein 13; zinc finger DHHC-type containing 13; zinc finger, DHHC domain containing 13; zinc finger, DHHC-type containing 13
Gene Aliases: 2410004E01Rik; C530010M18; fi22e09; HIP14L; HIP3RP; kojak; skc4; wu:fb06e01; wu:fc39g10; wu:fi22e09; ZDHHC13; zgc:101690
UniProt ID: (Human) Q8IUH4, (Mouse) Q9CWU2, (Zebrafish) A0A0R4IQZ2
Entrez Gene ID: (Horse) 100071954, (Human) 54503, (Bovine) 514176, (Rabbit) 100339374, (Dog) 613004, (Mouse) 243983, (Rat) 365252, (Guinea pig) 100724523, (Zebrafish) 494107
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.